

Palladium(II) chloride (≥59.5% Pd) ≥99.999% (metals basis) Pd ≥ 59.5%, Premion®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-10-1
:
Formula:PdCl₂
MDL Number:MFCD00003558
Merck Index:14,6990
Melting Pt:675 °C (dec.)
Density:4.0
EINECS:231-596-2
UN:3260
ADR:8,III
:177.33 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together