


Palladium(II) chloride 99.999% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-10-1
:
Formula:PdCl₂
MDL Number:MFCD00003558
Merck Index:13,07058
Melting Pt:678…680 °C (dec.)
Density:4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-596-2
UN:3260
ADR:8,III
:177.33 g/mol
:GHS07
:GHS05
:GHS09
Specification Test Results
- Appearance:Brown Powder
- Assay:59 to 61 % (Pd)
- Total trace metal impurities:≤0.001 %
Specification Test Results
Frequently Bought Together