
Palladium(II) bromide ≥99.99% (metals basis) Pd ≥ 39.5%, Premion®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13444-94-5
:
Formula:Br₂Pd
MDL Number:MFCD00011170
:266.23 g/mol
Melting Pt: Dec.
Density:5.173 g/cm³ (16 °C)
EINECS:236-588-2
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together