Human Recombinant GDF-15 (from E. coli)
About this item
Growth differentiation factor 15 (GDF-15) is a member of the transforming growth factor beta (TGF-β) family and is made by the placenta and cardiovascular tissues. GDF-15 regulates inflammatory and apoptotic pathways during cellular stress and injury. GDF-15 is emerging as a biomarker of early heart disease, such that increased levels of circulating GDF-15 are associated with an increased risk of developing heart failure. The GDF-15 D variant has a Histidine to an Aspartate substitution at amino acid position 7.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
5 Options Available Below
Product Details & Documents
Growth differentiation factor 15 (GDF-15) is a member of the transforming growth factor beta (TGF-β) family and is made by the placenta and cardiovascular tissues. GDF-15 regulates inflammatory and apoptotic pathways during cellular stress and injury. GDF-15 is emerging as a biomarker of early heart disease, such that increased levels of circulating GDF-15 are associated with an increased risk of developing heart failure. The GDF-15 D variant has a Histidine to an Aspartate substitution at amino acid position 7.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- SHBT100-171-5UG
- SHBT100-171-20UG
- SHBT100-171-100UG
- SHBT100-171-500UG
- SHBT100-171-1MG
- Protein/peptide name:
- GDF-15
- GDF-15
- GDF-15
- GDF-15
- GDF-15
- Protein synonyms:
- MIC-1|Prostate differentiation factor|Placental TGF-β
- MIC-1|Prostate differentiation factor|Placental TGF-β
- MIC-1|Prostate differentiation factor|Placental TGF-β
- MIC-1|Prostate differentiation factor|Placental TGF-β
- MIC-1|Prostate differentiation factor|Placental TGF-β
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Human
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- Q99988
- Q99988
- Q99988
- Q99988
- Q99988
- Sequence:
- MARNGDHCPL GPGRCCRLHT VRASLEDLGW ADWVLSPREV QVTMCIGACP SQFRAANMHA QIKTSLHRLK PDTVPAPCCV PASYNPMVLIQKTDTGVSLQ TYDDLLAKDC HCI
- MARNGDHCPL GPGRCCRLHT VRASLEDLGW ADWVLSPREV QVTMCIGACP SQFRAANMHA QIKTSLHRLK PDTVPAPCCV PASYNPMVLIQKTDTGVSLQ TYDDLLAKDC HCI
- MARNGDHCPL GPGRCCRLHT VRASLEDLGW ADWVLSPREV QVTMCIGACP SQFRAANMHA QIKTSLHRLK PDTVPAPCCV PASYNPMVLIQKTDTGVSLQ TYDDLLAKDC HCI
- MARNGDHCPL GPGRCCRLHT VRASLEDLGW ADWVLSPREV QVTMCIGACP SQFRAANMHA QIKTSLHRLK PDTVPAPCCV PASYNPMVLIQKTDTGVSLQ TYDDLLAKDC HCI
- MARNGDHCPL GPGRCCRLHT VRASLEDLGW ADWVLSPREV QVTMCIGACP SQFRAANMHA QIKTSLHRLK PDTVPAPCCV PASYNPMVLIQKTDTGVSLQ TYDDLLAKDC HCI
- Protease-free:
- N
- N
- N
- N
- N
- Animal-free:
- N
- N
- N
- N
- N
- Nuclease-free:
- N
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic acid (TFA)
- 0,1% Trifluoroacetic acid (TFA)
- 0,1% Trifluoroacetic acid (TFA)
- 0,1% Trifluoroacetic acid (TFA)
- Molecular weight:
- 12,4/24,8 kDa
- 12,4/24,8 kDa
- 12,4/24,8 kDa
- 12,4/24,8 kDa
- 12,4/24,8 kDa
- Reconstitution instructions:
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Storage conditions:
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- RT
- Tested applications:
- Bioassay
- Bioassay
- Bioassay
- Bioassay
- Bioassay
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.SHBT100-171-5UGProtein/peptide nameGDF-15Protein synonymsMIC-1|Prostate differentiation factor|Placental TGF-βProtein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDQ99988SequenceMARNGDHCPL GPGRCCRLHT VRASLEDLGW ADWVLSPREV QVTMCIGACP SQFRAANMHA QIKTSLHRLK PDTVPAPCCV PASYNPMVLIQKTDTGVSLQ TYDDLLAKDC HCIProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight12,4/24,8 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT100-171-20UGProtein/peptide nameGDF-15Protein synonymsMIC-1|Prostate differentiation factor|Placental TGF-βProtein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDQ99988SequenceMARNGDHCPL GPGRCCRLHT VRASLEDLGW ADWVLSPREV QVTMCIGACP SQFRAANMHA QIKTSLHRLK PDTVPAPCCV PASYNPMVLIQKTDTGVSLQ TYDDLLAKDC HCIProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight12,4/24,8 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT100-171-100UGProtein/peptide nameGDF-15Protein synonymsMIC-1|Prostate differentiation factor|Placental TGF-βProtein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDQ99988SequenceMARNGDHCPL GPGRCCRLHT VRASLEDLGW ADWVLSPREV QVTMCIGACP SQFRAANMHA QIKTSLHRLK PDTVPAPCCV PASYNPMVLIQKTDTGVSLQ TYDDLLAKDC HCIProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight12,4/24,8 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT100-171-500UGProtein/peptide nameGDF-15Protein synonymsMIC-1|Prostate differentiation factor|Placental TGF-βProtein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDQ99988SequenceMARNGDHCPL GPGRCCRLHT VRASLEDLGW ADWVLSPREV QVTMCIGACP SQFRAANMHA QIKTSLHRLK PDTVPAPCCV PASYNPMVLIQKTDTGVSLQ TYDDLLAKDC HCIProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight12,4/24,8 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT100-171-1MGProtein/peptide nameGDF-15Protein synonymsMIC-1|Prostate differentiation factor|Placental TGF-βProtein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDQ99988SequenceMARNGDHCPL GPGRCCRLHT VRASLEDLGW ADWVLSPREV QVTMCIGACP SQFRAANMHA QIKTSLHRLK PDTVPAPCCV PASYNPMVLIQKTDTGVSLQ TYDDLLAKDC HCIProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight12,4/24,8 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRTTested applicationsBioassay



