Orders LinkContactUsLinkComponent
Phase separating filters
Supplier:   PRAT DUMAS
Phase separating filters
Supplier:  PRAT DUMAS
Certificates
Phase separating filters
Phase separating filters
8 Options Available Below
Product Details & Documents
Hydrophobic paper for the fast separation of aqueous solutions from hydrofuge solvents.
Useful as well with organic solvents heavier or lighter than water replacing many times bulbs to be settled.
 : Filters supplied as flat in boxes with 100 filters or folded in 4 by 100.
Product Family Options
Product Information
  • Grade
    Size
    Availability
    Price
  • 108 H
    90 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 56,70 €
    56,70 €
    1 * 100 ST
       56,70 €
  • 108 H
    110 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 73,00 €
    73,00 €
    1 * 100 ST
       73,00 €
  • 108 H
    125 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 82,50 €
    82,50 €
    1 * 100 ST
       82,50 €
  • 108 H
    150 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 101,00 €
    101,00 €
    1 * 100 ST
       101,00 €
  • 108 H
    185 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 137,00 €
    137,00 €
    1 * 100 ST
       137,00 €
  • 108 H
    210 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 170,00 €
    170,00 €
    1 * 100 ST
       170,00 €
  • 108 H
    240 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 207,00 €
    207,00 €
    1 * 100 ST
       207,00 €
  • 108 H
    270 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 248,00 €
    248,00 €
    1 * 100 ST
       248,00 €
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.