Order Entry
Belgium
Orders LinkContactUsLinkComponent
Vanadium(V) oxide, Supelco®

Vanadium(V) oxide, Supelco®

Health
Irritant
Environment
Supplier:   Merck
Vanadium(V) oxide, Supelco®
Supplier:  Merck
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Vanadium pentoxide, di-Vanadium pentaoxide
  • CAS Number:1314-62-1
  • Formula:O₅V₂
  • MDL Number:MFCD00011457
  • Molecular Weight:181.88 g/mol
  • Merck Index:13,09987
  • Boiling Pt:1750 °C (1013 hPa)
  • Melting Pt:690 °C
  • Density:3.35 g/cm³ (20 °C)
  • EINECS:215-239-8
  • ADR:6.1,III
  •  :M-Clarity™ quality level = MQ200
  • UN:2862

Documents

Specifications for Product Family

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    690 °C
  • Molar Mass:
    181.88 g/mol
  • Bulk Density:
    400 - 600 kg/m3
  • Boiling Point:
    1750 °C (decomposition)
  • Density:
    3.36 g/cm3 (20 °C)
  • pH:
    4 (50 g/l, H2O, 20 °C) (slurry)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 690,00 €
    690,00 €
    1 * 250 g
       690,00 €

  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 2 420,00 €
    2 420,00 €
    1 * 1 kg
       2 420,00 €