Vanadium(V) oxide ≥99.99% (metals basis)
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99.99% CAS No.: 1314-62-1 Molecular Formula: V2O5 Formula Weight: 181.88 Density: 3.357 Appearance: Orange Odour: Odourless Melting Point: 690° Boiling Point: 1750° dec. Merck Reference: 14,9921 UN No.: 2862 MDL No.: MFCD00011457
Synonyms: Vanadium pentoxide, di-Vanadium pentaoxide
- CAS Number:1314-62-1
- Formula:O₅V₂
- MDL Number:MFCD00011457
- Molecular Weight:181.88 g/mol
- Merck Index:14,9921
- Boiling Pt:1750 °C (dec.)
- Melting Pt:690 °C
- Density:3.357
- Storage Temperature:Ambient
- EINECS:215-239-8
- UN:2862
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Purity: 99.99% CAS No.: 1314-62-1 Molecular Formula: V2O5 Formula Weight: 181.88 Density: 3.357 Appearance: Orange Odour: Odourless Melting Point: 690° Boiling Point: 1750° dec. Merck Reference: 14,9921 UN No.: 2862 MDL No.: MFCD00011457
Synonyms: Vanadium pentoxide, di-Vanadium pentaoxide
- CAS Number:1314-62-1
- Formula:O₅V₂
- MDL Number:MFCD00011457
- Molecular Weight:181.88 g/mol
- Merck Index:14,9921
- Boiling Pt:1750 °C (dec.)
- Melting Pt:690 °C
- Density:3.357
- Storage Temperature:Ambient
- EINECS:215-239-8
- UN:2862
- ADR:6.1,III



