Order Entry
Belgium
Orders LinkContactUsLinkComponent
Vanadium(V) oxide 99.6+%

Vanadium(V) oxide 99.6+%

Corrosive
Irritant
Health
Environment
Vanadium(V) oxide 99.6+%
Certificates
Vanadium(V) oxide 99.6+%

2 Options Available Below

Product Details & Documents
Synonyms:  Vanadium pentoxide, di-Vanadium pentaoxide
  • CAS Number:1314-62-1
  • Formula:O₅V₂
  • MDL Number:MFCD00011457
  • Molecular Weight:181.88 g/mol
  • Merck Index:13,09987
  • Boiling Pt:1750 °C (760 mmHg)
  • Melting Pt:690 °C
  • Density:3.35 g/cm³
  • Storage Temperature:Ambient
  • EINECS:215-239-8
  • UN:2862
  • ADR:6.1,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Yellow to orange-brown Powder
  • Assay:
    ≥99.60 %
  • Chromium (Cr):
    ≤100 ppm
  • Iron (Fe):
    ≤300 ppm
  • Potassium (K):
    ≤200 ppm
  • Silicon (Si):
    ≤200 ppm
  • Sodium (Na):
    ≤200 ppm
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 93,50 €
    93,50 €
    1 * 250 g
       93,50 €

  • Glass bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 275,00 €
    275,00 €
    1 * 1 kg
       275,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.