Order Entry
Belgium
ContactUsLinkComponent
Zirconium(IV) oxide ≥99% (metals basis excluding Hf), nanopowder
Irritant
Zirconium(IV) oxide ≥99% (metals basis excluding Hf), nanopowder
Catalog # 44886.14
CAS Number:  
Irritant
Zirconium(IV) oxide ≥99% (metals basis excluding Hf), nanopowder
Catalog # 44886.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :123.22 g/mol
Merck Index:14,10180
Boiling Pt:∼5000 °C
Melting Pt:∼2700 °C
Density:5.89
Storage Temperature:Ambient
EINECS:215-227-2
 :GHS07
CAS Number:1314-23-4
Formula:ZrO₂
MDL Number:MFCD00011310