Order Entry
Belgium
ContactUsLinkComponent
Titanium(IV) oxide ≥99%, GPR RECTAPUR®
Titanium(IV) oxide ≥99%, GPR RECTAPUR®
Catalog # 20732.298
Supplier:  VWR Chemicals
CAS Number:  
undefined
Titanium(IV) oxide ≥99%, GPR RECTAPUR®
Catalog # 20732.298
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Loss on ignition (800°C):
    Max. 0.5 %
  • As (Arsenic):
    Max. 5 ppm
  • Fe (Iron):
    Max. 0.02 %

Specification Test Results

About this item

 :
 :79.88 g/mol
Merck Index:13,09549
Boiling Pt:2900 °C (1013 hPa)
Melting Pt:1855 °C
Density:4.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:236-675-5
CAS Number:13463-67-7
 :1317-70-0
Formula:TiO₂
MDL Number:MFCD00011269