Order Entry
Belgium
ContactUsLinkComponent
 
Titanium(IV) oxide 99.0-100.5% (dried basis), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, E171, SAFC®
 
Titanium(IV) oxide 99.0-100.5% (dried basis), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, E171, SAFC®
 

 

  • Assay (cerimetric, calculated on dried substance):
    99.0 - 100.5 %
  • Identity:
    passes test
  • Appearance of solution:
    passes test
  • Substances soluble in water (Hotextract):
    ≤ 0.5 %
  • Substances soluble in water (Coldextract):
    ≤ 0.25 %
  • Substances soluble in dilute hydrochloric acid:
    ≤ 0.5 %
  • Acidity or alkalinity:
    passes test
  • Aluminium oxide and/or Silicon dioxide:
    ≤ 2.0 %
  • Heavy metals (as Pb):
    ≤ 0.002 %
  • Acid soluble barium (Ba):
    passes test
  • Acid soluble antimony (Sb):
    ≤ 0.0002 %
  • As (Arsenic):
    ≤ 0.0001 %
  • Cd (Cadmium):
    ≤ 0.00005 %
  • Fe (Iron):
    ≤ 0.02 %
  • Hg (Mercury):
    ≤ 0.0001 %
  • Pb (Lead):
    ≤ 0.0010 %
  • Sb (Antimony):
    ≤ 0.005 %
  • V (Vanadium)*:
    ≤ 0.0010 %
  • Zn (Zinc):
    ≤ 0.005 %
  • Residual solvents (Ph Eur/USP/ICH):
    excluded by the manufacturing process
  • Microbiological test (Total yeast and mould count (TYMC)):
    ≤ 100 CFU/g
  • Microbiological test (Total aerobic microbial count (TAMC)):
    ≤ 100 CFU/g
  • Particle Size (d10):
    ≥ 0.1 µm
  • Particle Size (d50):
    about 0.8 
  • Particle Size (d90):
    ≤ 2.5 µm
  • Loss on ignition (800 °C):
    ≤ 0.5 %
  • Loss on drying (105 °C; 3 h):
    ≤ 0.5 %

 

Frequently Bought Together