
Titanium(IV) oxide ≥99.9% (metals basis), high surface area
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13463-67-7
:1317-70-0
Formula:TiO₂
MDL Number:MFCD00011269
:79.88 g/mol
Merck Index:14,9472
Boiling Pt:2500…3000 °C
Melting Pt:1830…1850 °C
Density:4.26
EINECS:236-675-5
:GHS08
Storage Temperature:Ambient
Related Information
Frequently Bought Together