Power selector, WRT00116
Supplier: WINKLER
À propos de cet article
Plug-in power selector made of PS with switching mechanism.
- Rotary switch with bimetal element
- Signal light for controlling status indication
- Cable with grounding plug type
1 Options disponibles ci-dessous
Product Details & Documents
Plug-in power selector made of PS with switching mechanism.
- Rotary switch with bimetal element
- Signal light for controlling status indication
- Cable with grounding plug type
Spécifications de la famille de produits
Spécifications
- Catalog No:
- WINKWRT00116-XX013
- WINKWRT00116-XX0CH
- WINKWRT00116-XX0UK
- Modèle:
- WRT00116
- WRT00116
- WRT00116
- Exigence électrique:
- 230 V; 13 A (ohmic load); 50 Hz
- 230 V; 13 A (ohmic load); 50 Hz
- 230 V; 13 A (ohmic load); 50 Hz
- L×P×H:
- 120×56×57 mm
- 120×56×57 mm
- 120×56×57 mm
Spécifications
Product Family Options
Informations sur le produit
- DésignationModèleAvailabilityPrice
- Spécifications :
More
Specifications
Cat. No.WINKWRT00116-XX013ModèleWRT00116Exigence électrique230 V; 13 A (ohmic load); 50 HzL×P×H120×56×57 mm - Power selector, CH-plugWRT00116Discontinued-Spécifications :
More
Specifications
Cat. No.WINKWRT00116-XX0CHModèleWRT00116Exigence électrique230 V; 13 A (ohmic load); 50 HzL×P×H120×56×57 mm - Power selector, UK-plugWRT00116Discontinued-Spécifications :
More
Specifications
Cat. No.WINKWRT00116-XX0UKModèleWRT00116Exigence électrique230 V; 13 A (ohmic load); 50 HzL×P×H120×56×57 mm