Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Silver sulphate

Corrosive
Marketsource
Supplier:   Roth Carl
Silver sulphate
Supplier:  Roth Carl
Marketsource
Certificates
Silver sulphate

2 Options Available Below

Product Details & Documents
Synonyms:  Disilver sulphate
  • CAS Number:10294-26-5
  • Formula:Ag₂SO₄
  • MDL Number:MFCD00003407
  • Molecular Weight:311.8 g/mol
  • Merck Index:13,08603
  • Boiling Pt:1085 °C (1013 hPa)
  • Melting Pt:655 °C
  • Density:5.45 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:233-653-7
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

    Specifications:
    Marketsource
    Marketsource
  • 100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g

    Specifications:
    Marketsource
    Marketsource