Paraffin wax blended with synthetic polymers, Solidification point: 56-58°C, in pastille form, Histosec® embedding agent for histology, without DMSO, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:8002-74-2
:8012-95-1
:M-Clarity™ quality level = MQ400
Storage Temperature:Ambient
:1
Specification Test Results
- Solidification point (rotating thermometer method):56 - 58 °C
- Effect of heating (at 60 °C/100 g):clear melt within max. 20 hours
- Solubility in xylene (at 20 °C/0.1 g in 5 ml):clear solute within max. 20 hours
- Cutting results (4 µm slices):conforms
- Solubility of slices in xylene:50 seconds
Specification Test Results
Related Information
Frequently Bought Together