Copper(I) cyanide
About this item
CAS No.: 544-92-3 Molecular Formula: CCuN Formula Weight: 89.56 Physical Form: Powder Density: 2.92 Appearance: White to cream Odour: Characteristic Melting Point: 473° Solubility: Practically insoluble in water, alcohol. Soluble in NH4OH Beilstein Reference: 3587244 Merck Reference: 14,2661 UN No.: 1587 MDL No.: MFCD00010975
Electroplating Cu or Fe, polymerization catalyst
- CAS Number:544-92-3
- Formula:CCuN
- MDL Number:MFCD00010975
- Molecular Weight:89.56 g/mol
- Merck Index:14,2661
- Melting Pt:473 °C
- Density:2.92
- Storage Temperature:Ambient
- EINECS:208-883-6
- UN:1587
- ADR:6.1,II
3 Options Available Below
Product Details & Documents
CAS No.: 544-92-3 Molecular Formula: CCuN Formula Weight: 89.56 Physical Form: Powder Density: 2.92 Appearance: White to cream Odour: Characteristic Melting Point: 473° Solubility: Practically insoluble in water, alcohol. Soluble in NH4OH Beilstein Reference: 3587244 Merck Reference: 14,2661 UN No.: 1587 MDL No.: MFCD00010975
Electroplating Cu or Fe, polymerization catalyst
- CAS Number:544-92-3
- Formula:CCuN
- MDL Number:MFCD00010975
- Molecular Weight:89.56 g/mol
- Merck Index:14,2661
- Melting Pt:473 °C
- Density:2.92
- Storage Temperature:Ambient
- EINECS:208-883-6
- UN:1587
- ADR:6.1,II
Documents
Product Family Options
Product Information
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



