Orders LinkContactUsLinkComponent
Mop wipe systems, VertiKlean® MAX®
Mop wipe systems, VertiKlean® MAX®
Supplier:   CONTEC
Mop wipe systems, VertiKlean® MAX®
Supplier:  CONTEC
Certificates
Mop wipe systems, VertiKlean® MAX®
Mop wipe systems, VertiKlean® MAX®
3 Options Available Below
Product Details & Documents
The VertiKlean® MAX Mop Wipe Systems offer a quick and easy solution to mopping floors, walls, and other flat surfaces in any controlled environment. These versatile systems work with a variety of mop wipe covers, enabling the user to customize the cleaning task.
  • Stainless steel frame is chemical resistant
  • Easy to attach and remove mop covers
  • Variety of fabric options available
  • Removes contaminants easily from the environment
  • Slim profile allows easy access to confined spaces for efficient cleaning and disinfection
  • Thin, lightweight design makes it easy to clean floors, walls, and ceilings
These can be sterilised by most common methods including gamma irradiation, ethylene oxide, or autoclave (recommended cycle is 121 °C for 30 minutes). Pre-saturated or dry wipes are available to eliminate the need for a bucket and wringer.
Dry and pre-saturated single-use wipes fit over the VertiKlean® MAX foam mop head and QuickConnect frame to remove contamination such as powder, dirt, and dust from floors, walls, and ceilings. The VertiKlean® MAX Mop Wipe Systems provide excellent surface contact and conformity. It can be used to apply disinfectant solutions to surfaces when used with a bucket and wringer. Most importantly, when used for disinfection, the mop wipe covers entrap microorganisms, enabling them to be removed from the environment with each change of wipe cover.
Product Family Options
Product Information
  • Colour
    L×W
    Packed
    Availability
    Price
  • 115-3201
    Polyester quilted mop wipes, Quiltec®
    Polyester quilted mop wipes, Quiltec®
    White
    250×420 mm
    20 wipes/bag; 12 bags/case
    Log in for availability
    Log in for availability
    1 * 240 ST
    1 * 240 ST - 735,00 €
    735,00 €
    1 * 240 ST
       735,00 €
  • 115-3202
    Polyester quilted mop wipes, presaturated with 70% IPA and 30% DI water, Quiltec®
    Polyester quilted mop wipes, presaturated with 70% IPA and 30% DI water, Quiltec®
    White
    250×420 mm
    20 wipes/bag; 9 bags/case
    Log in for availability
    Log in for availability
    1 * 180 ST
    1 * 180 ST - 1 210,00 €
    1 210,00 €
    1 * 180 ST
       1 210,00 €
  • 115-1861
    MicroGenesis™ mop wipes, microfibre
    MicroGenesis™ mop wipes, microfibre
    White
    250×460 mm
    100 wipes/bag; 4 bags/case
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 815,00 €
    815,00 €
    1 * 400 ST
       815,00 €