Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Supelco® Organic Reference Standard, 2-Methylnaphthalene

2-Methylnaphthalene, Analytical standard, 250 mg
Catalog # 45796-250MG
Supplier:   Merck
Supelco® Organic Reference Standard, 2-Methylnaphthalene
Catalog # 45796-250MG
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, 2-Methylnaphthalene
Supelco® Organic Reference Standard, 2-Methylnaphthalene

2-Methylnaphthalene, Analytical standard, 250 mg

CAS Number:  
Price
50,60 €
Log in for availability
1 * 250 mg
Product Details & Documents
  • GHS signal: GHS07, GHS09

 : P261 - P264 - P273 - P301 + P312 - P302 + P352 - P305 + P351 + P338

 

Specifications

Specifications

  • Pk:
    250 mg
  • Standards:
    Analytical standard
  • Description:
    2-Methylnaphthalene
  • Cat. No.:
    45796-250MG
  • M-Clarity quality level:
    MQ100
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H315 - H319 - H335 - H411
  • MDL No.:
    MFCD00004118
  • Synonym(s):
    β-Methylnaphthalene
  • Molecular weight:
    142,20
  • CAS No.:
    91-57-6
  • Shipping conditions:
    STD

Specifications