Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

2-Naphthoic acid

Glass bottle for solids, 100 g
Catalog # MOLE48753954-100G
Marketsource
Supplier:   Molekula
2-Naphthoic acid
Catalog # MOLE48753954-100G
Supplier:  Molekula
Marketsource
CAS Number:  93-09-4
2-Naphthoic acid

Glass bottle for solids, 100 g

CAS Number:  93-09-4
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  β-Naphthoic acid
  • CAS Number:93-09-4
  • Formula:C₁₁H₈O₂
  • MDL Number:MFCD00004101
  • Molecular Weight:172.18 g/mol
  • Merck Index:12,06469
  • Boiling Pt:333 °C (1013 hPa)
  • Melting Pt:180…183 °C
  • Density:1.382 g/cm³ (20 °C)
  • EINECS:202-217-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications