Order Entry
Belgium
ContactUsLinkComponent
Ammonium cerium(IV) nitrate ≥98.5% ACS, Fluka™
Environment
Irritant
Corrosive
Oxidizing
Ammonium cerium(IV) nitrate ≥98.5% ACS, Fluka™
Catalog # 215473-50G
CAS Number:  
Environment
Irritant
Corrosive
Oxidizing
Ammonium cerium(IV) nitrate ≥98.5% ACS, Fluka™
Catalog # 215473-50G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Assay:
    Min. 98.5 %
  • Insoluble in acid:
    Max. 0.05 %
  • Iron (Fe):
    Max. 50 ppm
  • Chloride (Cl):
    Max. 100 ppm
  • Phosphate (PO₄):
    Max. 200 ppm

Specification Test Results

About this item

 :
 :548.22 g/mol
Merck Index:13,00509
Melting Pt:214 °C
Density:2.49 g/cm3
Storage Temperature:Ambient
EINECS:240-827-6
UN:3085
ADR:5.1,II
 :GHS09
 :GHS07
 :GHS05
 :GHS03
CAS Number:16774-21-3
Formula:(NH₄)₂Ce(NO₃)₆
MDL Number:MFCD00151121