


Ammonium cerium(IV) nitrate ≥95%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:16774-21-3
:
Formula:(NH₄)₂Ce(NO₃)₆
MDL Number:MFCD00151121
:548.22 g/mol
Merck Index:13,00509
Melting Pt:107…108 °C
Density:2.2 g/cm³ (20 °C)
EINECS:240-827-6
UN:1477
ADR:5.1,III
:GHS03
:GHS07
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 95 %
- Not precipitated by NH4OH (as SO4):Max. 0.2 %
- Cl (Chloride):Max. 0.02 %
Specification Test Results
Frequently Bought Together