

Ammonium cerium(IV) nitrate ≥98.5% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16774-21-3
:
Formula:(NH₄)₂Ce(NO₃)₆
MDL Number:MFCD00151121
:548.22 g/mol
Merck Index:14,1992
Melting Pt:107…108 °C
Density:2.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:240-827-6
UN:1477
ADR:5.1,II
:GHS03
:GHS07
Related Information
Frequently Bought Together