Order Entry
Belgium
Orders LinkContactUsLinkComponent
5-Nitroindole

5-Nitroindole

Glass bottle for solids, 5 g
Catalog # MOLEM20819045
Marketsource
Supplier:   Molekula
5-Nitroindole
Catalog # MOLEM20819045
Supplier:  Molekula
Marketsource
CAS Number:  6146-52-7
5-Nitroindole

Glass bottle for solids, 5 g

CAS Number:  6146-52-7
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  5-Nitro-1H-indole
  • CAS Number:6146-52-7
  • Formula:C₈H₆N₂O₂
  • MDL Number:MFCD00005673
  • Molecular Weight:162.15 g/mol
  • Melting Pt:140 °C
  • EINECS:228-153-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.