5-Nitroindole
Marketsource
Supplier: Thermo Scientific
Jump to:
About this item
Synonyms: 5-Nitro-1H-indole
- CAS Number:6146-52-7
- Formula:C₈H₆N₂O₂
- MDL Number:MFCD00005673
- Molecular Weight:162.15 g/mol
- Melting Pt:140 °C
- EINECS:228-153-0
2 Options Available Below
Product Details & Documents
Synonyms: 5-Nitro-1H-indole
- CAS Number:6146-52-7
- Formula:C₈H₆N₂O₂
- MDL Number:MFCD00005673
- Molecular Weight:162.15 g/mol
- Melting Pt:140 °C
- EINECS:228-153-0
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



