Order Entry
Belgium
Orders LinkContactUsLinkComponent
4-Fluoroindole

4-Fluoroindole

Glass bottle for solids, 1 g
Catalog # MOLE39257867-1G
Irritant
Marketsource
Supplier:   Molekula
4-Fluoroindole
Catalog # MOLE39257867-1G
Supplier:  Molekula
Marketsource
CAS Number:  387-43-9
4-Fluoroindole

Glass bottle for solids, 1 g

CAS Number:  387-43-9
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  4-Fluoro-1H-indole
  • CAS Number:387-43-9
  • Formula:C₈H₆FN
  • MDL Number:MFCD00055992
  • Molecular Weight:135.14 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.