Ninhydrin ACS
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
- CAS Number:485-47-2
- Formula:C₉H₆O₄
- MDL Number:MFCD00003791
- Molecular Weight:178.14 g/mol
- Merck Index:13,06582
- Boiling Pt:351 °C (1013 hPa)
- Melting Pt:250 °C (dec.)
- Density:0,862 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:207-618-1
4 Options Available Below
Product Details & Documents
Synonyms: Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
- CAS Number:485-47-2
- Formula:C₉H₆O₄
- MDL Number:MFCD00003791
- Molecular Weight:178.14 g/mol
- Merck Index:13,06582
- Boiling Pt:351 °C (1013 hPa)
- Melting Pt:250 °C (dec.)
- Density:0,862 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:207-618-1
Documents
ACRO41572
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White to light brown Crystals
- Infrared spectrum:Conforms
- Melting point:Passes test
- Solubility:Passes test
- Identification:Passes test
- Sensitivity:(to amino acids) Passes test



