Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ninhydrin for TLC

Ninhydrin for TLC

Spray can, 240 mL
Catalog # ACRO327022400
Flammable
Corrosive
Irritant
Health
Ninhydrin for TLC
Catalog # ACRO327022400
CAS Number:  485-47-2
Ninhydrin for TLC
Ninhydrin for TLC

Spray can, 240 mL

CAS Number:  485-47-2
Price
102,00 €
Log in for availability
1 * 240 mL
Product Details & Documents

Ozone friendly ready, to use spray for TLC

Synonyms:  Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
  • CAS Number:485-47-2
  • Formula:C₉H₆O₄
  • MDL Number:MFCD00003791
  • Molecular Weight:178.14 g/mol
  • Merck Index:13,06582
  • Boiling Pt:351 °C (1013 hPa)
  • Melting Pt:250…258 °C
  • Density:0,862 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-618-1
  • UN:1950

Documents

Specifications

Specification Test Results

  • Additional info:
    TLC spray reagent at 0.5 % in 1-Butanol

Specifications

  • Pack type:
    Spray can
  • Pk:
    240 mL

Specifications