Dimethyl sulphoxide, anhydrous (max. 0.005% H₂O) ≥99.8%
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:67-68-5
:
Formula:(CH₃)₂SO
MDL Number:MFCD00002089
:78.14 g/mol
Merck Index:13,03285
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18.5 °C
Density:1.101 g/cm³ (20 °C)
:Filtered through a 0.2 µm filter, packaged under nitrogen.
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:200-664-3
REACH:01-2119431362-50
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 99.8 %
- Acidity:Max. 0.0005 meq/g
- Residue on evaporation:Max. 0.0005 %
- Water:Max. 50 ppm
Specification Test Results
Frequently Bought Together