Order Entry
Belgium
ContactUsLinkComponent
Dimethyl sulphoxide, dried (max. 0.025% H₂O) for DNA and peptides synthesis
Dimethyl sulphoxide, dried (max. 0.025% H₂O) for DNA and peptides synthesis
Supplier:  Merck
   Sustainable Options Available
Certificates
undefined
Dimethyl sulphoxide, dried (max. 0.025% H₂O) for DNA and peptides synthesis
Supplier:  Merck
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

Specification Test Results

  • Solubility:
    1000 g/l (20 °C)
  • Melting Point:
    18.4 °C
  • Molar Mass:
    78.1 g/mol
  • Boiling Point:
    189 °C (1013 hPa)
  • Vapor Pressure:
    0.6 hPa (20 °C)
  • Flash Point:
    87 °C
  • Refractive Index:
    1.48 (20 °C)
  • Explosion Limit:
    1.8 - 63.0 %(V)
  • Density:
    1.10 g/cm3 (20 °C)
  • Ignition Point:
    301 °C
  • Saturation concentration:
    8.0 g/m3 (20 °C)
  • Viscosity kinematic:
    2.14 mm2/s

Specification Test Results

Related Information

Frequently Bought Together