Order Entry
Belgium
ContactUsLinkComponent
Dimethyl sulphoxide ≥99%, GPR RECTAPUR®
Dimethyl sulphoxide ≥99%, GPR RECTAPUR®
Catalog # 282164K
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  
undefined
Dimethyl sulphoxide ≥99%, GPR RECTAPUR®
Catalog # 282164K
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

Specification Test Results

  • IR Spectrum:
    Passes test
  • Assay:
    Min. 99 %
  • Evaporation residue:
    Max. 50 ppm
  • Water:
    Max. 0.05 %
  • Cu (Copper):
    Max. 5 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Pb (Lead):
    Max. 5 ppm

Specification Test Results

About this item

 :
 :78.14 g/mol
Merck Index:13,03285
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18.5 °C
Density:1.101 g/cm³ (20 °C)
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:200-664-3
REACH:01-2119431362-50
CAS Number:67-68-5
Formula:(CH₃)₂SO
MDL Number:MFCD00002089