Order Entry
Belgium
ContactUsLinkComponent
 
Dimethyl sulphoxide, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
  7093.1000
 :  Avantor
   Safer Human and Environmental Health
 :  
undefined
Dimethyl sulphoxide, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
  7093.1000
 :  Avantor
   Safer Human and Environmental Health
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

 

 

  • Assay:
    min. 99.5 %
  • Color (APHA):
    max. 10
  • Residue after Evaporation:
    max. 0.005 %
  • Water (H2O):
    max. 0.05 %
  • at 270 nm:
    max. 1.0
  • at 275 nm:
    max. 0.40
  • at 280 nm:
    max. 0.30
  • at 300 nm:
    max. 0.10
  • at 335 nm:
    max. 0.03
  • at 400 nm:
    max. 0.01

 

 

 :
 :78.14 g/mol
 :13,03285
 :189 °C (1013 hPa)
 :18.5 °C
 :1.101 g/cm³ (20 °C)
 :87 °C
 :Ambient
 :200-664-3
 :67-68-5
 :(CH₃)₂SO
 :MFCD00002089