Ammonium sulphate
Supplier: Roth Carl
Certificates
About this item
CAS Number:7783-20-2
:
Formula:(NH₄)₂SO₄
MDL Number:MFCD00003391
:132.14 g/mol
Merck Index:13,00559
Melting Pt:336…339 °C
Density:1.78 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-984-1
REACH:01-2119455044-46
Frequently Bought Together