Order Entry
Belgium
ContactUsLinkComponent
Ammonium sulphate
Ammonium sulphate
Catalog # ROTH9212.1
Supplier:  Roth Carl
CAS Number:  
Ammonium sulphate
Catalog # ROTH9212.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 kg

Specifications

About this item

 :
 :132.14 g/mol
Merck Index:13,00559
Melting Pt:336…339 °C
Density:1.78 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-984-1
REACH:01-2119455044-46
CAS Number:7783-20-2
Formula:(NH₄)₂SO₄
MDL Number:MFCD00003391