Order Entry
Belgium
ContactUsLinkComponent
Ammonium sulphate ≥99%, GPR RECTAPUR®
Ammonium sulphate ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Ammonium sulphate ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Ignition residue (SO4):
    Max. 0.02 %
  • Cl (Chloride):
    Max. 20 ppm
  • NO3 (Nitrate):
    Max. 50 ppm
  • Fe (Iron):
    Max. 10 ppm

Specification Test Results

Frequently Bought Together