Order Entry
Belgium
ContactUsLinkComponent
Ammonium sulphate, Suprapur® for trace analysis, Supelco®
Ammonium sulphate, Suprapur® for trace analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Ammonium sulphate, Suprapur® for trace analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    754 g/l (20 °C)
  • Molar Mass:
    132.14 g/mol
  • Bulk Density:
    850 kg/m3
  • Density:
    1.77 g/cm3 (20 °C)
  • pH:
    5 (100 g/l, H2O)

Specification Test Results

Related Information

Frequently Bought Together