Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ammonium sulphate
Ammonium sulphate, Millipore®
Ammonium sulphate, Millipore®
Ammonium sulphate, Millipore®
168356-1
:
Merck Millipore (Calbiochem)
:
Ammonium sulphate, Millipore®
168356-1
:
Merck Millipore (Calbiochem)
:
Pk:
1 kg
Seq:
4024
:
:
132.14 g/mol
:
13,00559
:
336…339 °C
:
1.78 g/cm³ (20 °C)
:
Ambient
:
231-984-1
:
01-2119455044-46
:
7783-20-2
:
(NH₄)₂SO₄
:
MFCD00003391