Order Entry
Belgium
ContactUsLinkComponent
Ammonium sulphate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Ammonium sulphate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
Ammonium sulphate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    min. 99.0 %
  • Arsenic (As):
    max. 0.3 ppm
  • Chloride (Cl):
    max. 3 ppm
  • Iron (Fe):
    max. 5 ppm
  • Heavy Metals (as Pb):
    max. 5 ppm
  • Insoluble Matter:
    max. 0.01 %
  • Nitrate (NO3):
    max. 5 ppm
  • pH of 5% Solution at 25°C:
    5.0 - 6.0
  • Phosphate (PO4):
    max. 5 ppm
  • Residue after Ignition:
    max. 0.01 %

Specification Test Results

Related Information

Frequently Bought Together