Order Entry
Belgium
ContactUsLinkComponent
Ammonium sulphate, granules FCC, J.T.Baker®
Ammonium sulphate, granules FCC, J.T.Baker®
Catalog # 0800-05
Supplier:  Avantor
CAS Number:  
undefined
Ammonium sulphate, granules FCC, J.T.Baker®
Catalog # 0800-05
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

Specification Test Results

  • Bulk Food Chemical:
    99.0 - 100.5 %
  • FCC - Assay:
    99.0 - 100.5 %
  • FCC - Identification:
    Passes Test
  • FCC - Lead (Pb):
    ≤3 mg/kg
  • FCC - Residue on Ignition:
    ≤0.25 %
  • FCC - Selenium (Se):
    ≤0.003 %

Specification Test Results

About this item

 :
 :132.14 g/mol
Merck Index:13,00559
Melting Pt:336…339 °C
Density:1.78 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-984-1
REACH:01-2119455044-46
CAS Number:7783-20-2
Formula:(NH₄)₂SO₄
MDL Number:MFCD00003391