Order Entry
Belgium
ContactUsLinkComponent
Ammonium sulphate 99.0-100.5%, granular FCC, ACS, J.T.Baker®
Health
Ammonium sulphate 99.0-100.5%, granular FCC, ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Health
Ammonium sulphate 99.0-100.5%, granular FCC, ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    99.0 - 100.5%
  • Food GMP Manufactured Product:
    ≤0.005%
  • Meets F.C.C. Requirements:
    ≤0.005%
  • CAUTION: For Manufacturing, processing or repackaging:
    5.0 - 6.0
  • Bulk Food Chemical:
    ≤0.001%
  • ACS - Assay ((NH₄)₂SO₄):
    99.0 - 100.5 %
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Residue on Ignition (ACS):
    ≤0.005 %
  • ACS - pH of 5% Solution at 25°C:
    5.0 - 6.0
  • ACS - Nitrate (NO₃):
    ≤0.001 %
  • Chloride (Cl):
    ≤3 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm
  • Phosphate (PO₄):
    ≤3 ppm
  • Heavy Metals (as Pb):
    ≤3 ppm
  • FCC - Assay ((NH₄)₂SO₄)(F.C.C.):
    99.0 - 100.5 %
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Residue on Ignition (FCC):
    ≤0.25 %
  • FCC - Lead (Pb):
    ≤3 mg/kg
  • FCC - Selenium (Se):
    ≤0.003 %

Specification Test Results

Related Information

Frequently Bought Together