
Ammonium hydrogen carbonate 99.0-101.0% (by acidimetry), EMPROVE® ESSENTIAL Ph. Eur., BP, E503, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:1066-33-7
:
Formula:NH₄HCO₃
MDL Number:MFCD00012138
:79.06 g/mol
Merck Index:13,00497
Melting Pt:35 °C
Density:1.57 g/cm³ (20 °C)
EINECS:213-911-5
:GHS07
Specification Test Results
- Assay (Acidimetric):99.0 - 101.0%
- Identity:Passes Test
- pH-Value (5 %; Water):7.5 - 8.1
- Chloride (Cl):≤0.001%
- Sulfate (SO₄):≤0.0030%
- Heavy Metals (as Pb):≤0.001%
- As (Arsenic):≤0.0002%
- Fe (Iron):≤0.0005%
- Hg (Mercury):≤0.0001%
- Pb (Lead):≤0.0002%
- Non-Volatile Matter:≤0.0500%
- Residual Solvents (ICH Q3C):Excluded by Manufacturing Process
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERP1.01131.5000_TDS_EN ProductPage_101131
Product specification
msds_101131
SDS
Frequently Bought Together