Order Entry
Belgium
ContactUsLinkComponent
 
Irritant
Ammonium chloride ≥99.9995% (metals basis), TraceSELECT™ for trace analysis, Fluka™
Catalog # 09725-25G
CAS Number:  
undefined
Irritant
Ammonium chloride ≥99.9995% (metals basis), TraceSELECT™ for trace analysis, Fluka™
Catalog # 09725-25G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Silver (Ag):
    Max. 0.01 ppm
  • Aluminium (Al):
    Max. 0.05 ppm
  • Arsenic (As):
    Max. 0.01 ppm
  • Gold (Au):
    Max. 0.01 ppm
  • Barium (Ba):
    Max. 0.1 ppm
  • Beryllium (Be):
    Max. 0.01 ppm
  • Bismuth (Bi):
    Max. 0.05 ppm
  • Calcium (Ca):
    Max. 0.5 ppm
  • Cadmium (Cd):
    Max. 0.01 ppm
  • Cerium (Ce):
    Max. 0.01 ppm
  • Cobalt (Co):
    Max. 0.01 ppm
  • Chromium (Cr):
    Max. 0.1 ppm
  • Cesium (Cs):
    Max. 0.1 ppm
  • Copper (Cu):
    Max. 0.01 ppm
  • Dysprosium (Dy):
    Max. 0.01 ppm
  • Erbium (Er):
    Max. 0.01 ppm
  • Europium (Eu):
    Max. 0.01 ppm
  • Iron (Fe):
    Max. 0.1 ppm
  • Gallium (Ga):
    Max. 0.05 ppm
  • Gadolinium (Gd):
    Max. 0.01 ppm
  • Germanium (Ge):
    Max. 0.05 ppm
  • Hafnium (Hf):
    Max. 0.01 ppm
  • Mercury (Hg):
    Max. 1 ppb
  • Holmium (Ho):
    Max. 0.01 ppm
  • Indium (In):
    Max. 0.01 ppm
  • Iridium (Ir):
    Max. 0.01 ppm
  • Potassium (K):
    Max. 0.5 ppm
  • Lanthanum (La):
    Max. 0.01 ppm
  • Lithium (Li):
    Max. 0.05 ppm
  • Lutetium (Lu):
    Max. 0.01 ppm
  • Magnesium (Mg):
    Max. 0.05 ppm
  • Manganese (Mn):
    Max. 0.01 ppm
  • Molybdenum (Mo):
    Max. 0.05 ppm
  • Sodium (Na):
    Max. 0.5 ppm
  • Niobium (nb):
    Max. 0.01 ppm
  • Neodymium (nd):
    Max. 0.01 ppm
  • Nickel (Ni):
    Max. 0.01 ppm
  • Lead (Pb):
    Max. 0.01 ppm
  • Palladium (Pd):
    Max. 0.01 ppm
  • Praseodymium (Pr):
    Max. 0.01 ppm
  • Platinum (Pt):
    Max. 0.1 ppm
  • Rubidium (Rb):
    Max. 0.01 ppm
  • Rhenium (Re):
    Max. 0.01 ppm
  • Rhodium (Rh):
    Max. 0.01 ppm
  • Ruthenium (Ru):
    Max. 0.01 ppm
  • Antimony (Sb):
    Max. 0.01 ppm
  • Scandium (Sc):
    Max. 0.05 ppm
  • Selenium (Se):
    Max. 0.05 ppm
  • Samarium (Sm):
    Max. 0.01 ppm
  • Tin (Sn):
    Max. 0.05 ppm
  • Strontium (Sr):
    Max. 0.1 ppm
  • Tantalum (Ta):
    Max. 0.01 ppm
  • Terbium (Tb):
    Max. 0.01 ppm
  • Tellurium (Te):
    Max. 0.01 ppm
  • Thorium (Th):
    Max. 0.01 ppm
  • Titanium (Ti):
    Max. 0.05 ppm
  • Thallium (Tl):
    Max. 0.01 ppm
  • Thulium (Tm):
    Max. 0.01 ppm
  • Uran (U):
    Max. 0.01 ppm
  • Vanadium (V):
    Max. 0.05 ppm
  • Tungsten (W):
    Max. 0.01 ppm
  • Yttrium (Y):
    Max. 0.05 ppm
  • Ytterbium (Yb):
    Max. 0.01 ppm
  • Zinc (Zn):
    Max. 0.01 ppm
  • Zirconium (Zr):
    Max. 0.01 ppm
  • Nitrate (NO₃):
    Max. 5 ppm
  • Phosphate (PO₄):
    Max. 2 ppm
  • Sulfate (SO₄):
    Max. 20 ppm

Specification Test Results

About this item

 :
 :53.49 g/mol
Merck Index:13,00510
Melting Pt:338 °C (dec.)
Density:1.536 g/cm³ (20 °C)
EINECS:235-186-4
 :GHS07
Storage Temperature:Ambient
CAS Number:12125-02-9
Formula:NH₄Cl
MDL Number:MFCD00011420