


Benzethonium chloride, anhydrous ≥95%, TECHNICAL
Catalog # 21798.235
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Assay:Min. 95 %
Specification Test Results
About this item
:
:448.09 g/mol
Merck Index:13,01074
Melting Pt:164…166 °C
Storage Temperature:Ambient
EINECS:204-479-9
UN:3077
ADR:9,III
:GHS07
:GHS05
:GHS09
CAS Number:121-54-0
Formula:C₂₇H₄₂ClNO₂
MDL Number:MFCD00011742