Tetrabutylammonium hydrogen sulphate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:32503-27-8
:
Formula:(C₄H₉)₄NHSO₄
MDL Number:MFCD00011637
:339.54 g/mol
Melting Pt:169…172 °C
Density:1.01 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ200
EINECS:251-068-5
:GHS07
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:168 - 174 °C
- Molar Mass:339.54 g/mol
- Bulk Density:650 kg/m3
- pH:1 - 2 (100 g/l, H2O, 20 °C)
Specification Test Results
Related Information
Frequently Bought Together