
Tetrabutylammonium hydrogen sulphate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:32503-27-8
:
Formula:(C₄H₉)₄NHSO₄
MDL Number:MFCD00011637
:339.54 g/mol
Melting Pt:168…172 °C
Density:1,01 g/cm³ (25 °C)
EINECS:251-068-5
:GHS07
Specification Test Results
- Appearance:White to off-white Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:168 to 173 °C
- Titration with NaOH:≥97.5 %
Specification Test Results
Frequently Bought Together