Choline chloride
Glass bottle for solids, 1 kg
Catalog # A0785.1000
Supplier: PanReac AppliChem
Jump to:
Product Details & Documents
Synonyms: 2-hydroxy-N,N,N-trimethylethanaminium chloride
- CAS Number:67-48-1
- Formula:HOH₂CCH₂N(CH₃)₃Cl
- MDL Number:MFCD00011721
- Molecular Weight:139.62 g/mol
- Merck Index:13,02226
- Melting Pt:244…247 °C
- Density:∼1.205 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-655-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



