Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 10 mol/l (10 N) volumetric solution
Corrosive
Sodium hydroxide 10 mol/l (10 N) volumetric solution
Catalog # APPC183508.1211
Supplier:  PanReac AppliChem
CAS Number:  
Corrosive
Sodium hydroxide 10 mol/l (10 N) volumetric solution
Catalog # APPC183508.1211
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer at 20 °C:
    0.99 - 1.01
  • Uncertainty:
    See certificate
  • Traceability:
    NIST

Specification Test Results

About this item

 :
 :40 g/mol
Density:1.331 kg/l
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548