Order Entry
Belgium
ContactUsLinkComponent
DENARASE® highly active endonuclease from Serratia marcescens
DENARASE® highly active endonuclease from Serratia marcescens
Supplier:  C LECTA
Certificates
undefined
DENARASE® highly active endonuclease from Serratia marcescens
Supplier:  C LECTA
Certificates

Specifications

  • Species:
    Serratia marcescens
  • Storage conditions:
    Store at −20 °C
  • Enzyme name:
    DENARASE
  • Enzyme activity:
    >250 U/µl
  • Purity:
    ≥99%
  • Molecular weight:
    27 kDa (per monomer)

Specifications

Frequently Bought Together