Triethyl phosphite ≥97.0% (by GC) for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:122-52-1
:
Formula:C₆H₁₅O₃P
MDL Number:MFCD00009084
Boiling Pt:156 °C (1013 hPa)
Melting Pt:51 °C
Density:0.9643 g/cm³ (20 °C)
Flash Pt:52 °C
EINECS:204-552-5
UN:2323
ADR:3,III
:M-Clarity™ quality level = MQ200
:166.16 g/mol
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Melting Point:⁻112 °C
- Molar Mass:166.16 g/mol
- Boiling Point:156 - 158 °C (1013 hPa)
- Vapor Pressure:- 6 hPa (20 °C)
- Flash Point:52 °C
- Refractive Index:1,4133 (20 °C, 589 nm)
- Explosion Limit:3,75 - 42,5 %(V)
- Density:0,956 g/cm³ (20 °C)
- pH:(H2O) not applicable
- Ignition Point:250 °C
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Specification Test Results
Frequently Bought Together