Orders LinkContactUsLinkComponent
Colour coded inserts, CapInsert™
Colour coded inserts, CapInsert™
Supplier:   Simport Scientific
Colour coded inserts, CapInsert™
Supplier:  Simport Scientific
Certificates
Colour coded inserts, CapInsert™
Colour coded inserts, CapInsert™
5 Options Available Below
Product Details & Documents
These colour coded inserts could be applied to the tube closure, which eliminates jeopardising the integrity of your sample by transferring it to another vial.
  • Manually press fit and locks into place on top of closure
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • Blue
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 21,80 €
    21,80 €
    1 * 200 ST
       21,80 €
  • Green
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 21,90 €
    21,90 €
    1 * 200 ST
       21,90 €
  • Purple
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 21,90 €
    21,90 €
    1 * 200 ST
       21,90 €
  • Red
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 21,80 €
    21,80 €
    1 * 200 ST
       21,80 €
  • Yellow
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 21,80 €
    21,80 €
    1 * 200 ST
       21,80 €