


Tributyltin chloride ≥96%
STRE93-5056-50G
:
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
:
:325.51 g/mol
:172 °C (5 mmHg)
:–9 °C
:1.207 (20°)
:210 °F
:215-958-7
:2788
:6.1,III
:GHS06
:GHS08
:GHS07
:1461-22-9
:C₁₂H₂₇ClSn
:MFCD00000521